Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ G418 Sulfate

Catalog No. 5092900005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-929-00005 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-929-00005 Supplier MilliporeSigma™ Supplier No. 5.09290.0005

missing translation for 'validMainframeMsg'

G418 is an aminoglycoside antibiotic related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis.

Aminoglycoside related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis. Toxic to bacteria, yeast, protozoans, helminths, higher plant, and mammalian cells. Widely used in the selection of eukaryotic expression vectors carrying the bacterial
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15-30°C
pH 4 to 6
Purity or Quality Grade ≥90% by HPLC
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.