Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Oxytetracycline, Hydrochloride

Catalog No. 500105100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-010-5100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-010-5100GM Supplier MilliporeSigma™ Supplier No. 500105100GM

missing translation for 'validMainframeMsg'

An antimicrobial agent

A member of the tetracycline family that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2-8°C
Purity or Quality Grade ≥95% by HPLC
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.