Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PANSORBIN™ Cells

Catalog No. 5078585GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-785-85GM 5 g
50-785-81GM 1 g
50-785-810GM 10 g
50-785-850GM 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-785-85GM Supplier MilliporeSigma™ Supplier No. 5078585GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For mitogenic stimulation of B lyphocytes and for immunoprecipitation

PANSORBIN cells are useful as a solid-phase IgG-binding reagent due to the high-affinity interaction of protein A with the Fc domain of IgG. They work better when the antibody is human (IgG1, IgG2, IgG4), rabbit IgG (all isotypes), or mouse (IgG2a, IgG2b, IgG3).

  • Activity: ≥2.0 mg human IgG/100mg cells
  • Form: Homogeneous, milky suspension
  • Formulation: Supplied as a 10% suspension of Staphylococcus aureus cells in PBS, 0.1% NaN3, pH 7.2
  • Contaminants: Viable cells: ≤2500 cells/mL
  • Storage: Store at 2° to 8°C. Do not freeze

TRUSTED_SUSTAINABILITY

Specifications

Product Type PANSORBIN Cells
Content And Storage 2°C to 8°C
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.