Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PANSORBIN™ Cells, Lyophilized

Catalog No. 5078621GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-786-21GM 1 g
50-786-25GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-786-21GM Supplier MilliporeSigma™ Supplier No. 5078621GM
Add to Cart
Add to Cart

Lyophilized cells for enzyme immunoassay, immunoprecipitation and radioimmunoassay

PANSORBIN cells are formalin-fixed and heat-killed Staphylococcus aureus cells bearing protein A that bind to the Fc portion of human IgG subclasses 1, 2, and 4, and to IgG from other species.

  • Form: Lyophilized
  • Formulation: Lyophilized from PBS, 0.1% NaN3
  • Reconstitution: Reconstitute in sterile distilled H2O
  • Soluble in sterile dH2O
  • Applications: Enzyme Immunoassay, Immunoprecipitation, Radioimmunoassay
  • Storage: Store at 2° to °C

TRUSTED_SUSTAINABILITY

Specifications

Product Type PANSORBIN Cells
Content And Storage 2°C to 8°C
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.