Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PANSORBIN™ Cells, Standardized

Catalog No. 50786150ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-786-150ML 50 mL
50-786-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-786-150ML Supplier MilliporeSigma™ Supplier No. 50786150ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For mitogenic stimulation of B lyphocytes and for immunoprecipitation

PANSORBIN cells are useful as a solid-phase IgG-binding reagent due to the high-affinity interaction of protein A with the Fc domain of IgG. They work better when the antibody is human (IgG1, IgG2, IgG4), rabbit IgG (all isotypes), or mouse (IgG2a, IgG2b, IgG3).

  • Activity: Binding capacity: ≥2mg/mL
  • Form: Liquid
  • Formulation: Supplied as a 7.5 to 15% (w/v) Staphylococcus aureus cell suspension in PBS, 0.1% sodium azide, pH 7.2
  • Preservative: 0.1% sodium azide
  • Storage: Store at 2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Product Type PANSORBIN Cells
Content And Storage 2°C to 8°C
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.