Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Penciclovir

Catalog No. 516090100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-609-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-609-0100MG Supplier MilliporeSigma™ Supplier No. 516090100MG

A carba analog of Ganciclovir active against several herpes viruses, induces apoptosis effectively without exerting much genotoxicity

A carba analog of Ganciclovir (Cat. No.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20° C
Purity or Quality Grade ≥98% by HPLC;
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.