Learn More
MilliporeSigma™ Calbiochem™ Penicillin G, Potassium Salt
Catalog No. 516125MU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mU
Catalog No. Quantity
51-612-5MU 25 mU
1 options
Catalog No. 51-612-5MU Supplier MilliporeSigma™ Supplier No. 516125MU
Antibiotic effective against Gram-positive bacteria. Blocks cell wall formation in growing cells, resulting in accumulation of uracil nucleotides of muramic acid peptides.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2-8°C
Antibiotic Spectrum Gram-Positive Bacteria
Quantity 25 mU