Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Polyvinylpyrrolidone

Catalog No. 5295100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-951-00GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-951-00GM Supplier MilliporeSigma™ Supplier No. 5295100GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polymer used as a pharmaceutical aid, complexing agent and solubilizer

Polymer used as a pharmaceutical aid, complexing agent, and a solubilizer.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Product Type Polyvinylpyrrolidone
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.