Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Radicicol, Diheterospora chlamydosporia

Catalog No. 553400500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-340-0500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-340-0500UG Supplier MilliporeSigma™ Supplier No. 553400500UG
Add to Cart
Add to Cart

A cell-permeable, ATP-site binding, and irreversible antifungal macrocyclic lactone antibiotic that acts as a protein tyrosine kinase inhibitor

Inhibits p60v-src kinase activity (IC50 = 0.27 nM). Inhibits the expression of COX-2 (IC50 = 27 nM) without affecting COX-1 expression in LPS-stimulated macrophages. Disrupts K-Ras-activated signaling pathways by selectively depleting Raf kinase. Inhibits tyrosine phosphorylation of p53/56lyn in LPS-stimulated macrophages. Suppresses NIH/3T3 cell transformation by diverse oncogenes such as src, ras, and mos in part by blocking the key signal transduction intermediates such as MAP kinase and GAP-associated p62. Also exhibits anti-angiogenic activity in vivo. Inhibits Wnt-5A expression in dermal papilla cells (IC50 = 190 nM).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2-8°C
Purity or Quality Grade ≥98% by HPLC
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.