Learn More
MilliporeSigma™ Calbiochem™ Reagent Grade TCEP, Hydrochloride Reducing Agent
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| For Use With (Application) | Reducing Agent |
| Content And Storage | Store at 2°C to 8°C Protect from moisture |
| Form | Solid |
| Quantity | 5 g |
| Synonym | tris(2-Carboxyethyl)phosphine, HCl |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.