Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Recombinant Human Epidermal Growth Factor

Catalog No. 324831200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-483-1200UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-483-1200UG Supplier MilliporeSigma™ Supplier No. 324831200UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

expressed in E. coli

Recombinant, human epidermal growth factor (54 amino acids) expressed in E. coli. Identical to human EGF except for an additional N-terminal methionine. Human EGF is 70% homologous to mouse EGF and has been shown to have similar biological activity. Mitogenic for a wide range of ectoderm- and endoderm-derived cells from tissue such as breast, cornea, epidermis, dermis, liver, pancreas, nerve, amnion, and adrenal medulla. Acts as a survival factor in preventing apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Formulation Lyophilized from sterile-filtered PBS
Quantity 20 μg
Source E. coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.