Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Streptavidin, Alkaline Phosphatase Conjugate

Catalog No. 1897322ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-973-22ML 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-973-22ML Supplier MilliporeSigma™ Supplier No. 1897322ML
Only null left
Add to Cart
Add to Cart

For ELISA, Western Blotting, Immunohistochemistry

  • Specific for biotin, biotinylated proteins, or biotinylated proteins
  • Form: Liquid
  • Formulation: In proprietary buffer
  • Preservative: ≤0.1% sodium azide
TRUSTED_SUSTAINABILITY

Specifications

Description Streptavidin, Alkaline Phosphatase Conjugate
Quantity 2 mL
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.