Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calcitonin, Salmon, Synthetic, Peptide

Catalog No. 0523240125M Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-232-40125M 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-232-40125M Supplier MilliporeSigma™ Supplier No. 0523240125MG
Add to Cart
Add to Cart
Synthetic, salmon calcitonin. A 32 amino acid residue peptide that is shown to stimulate bone formation and inhibit bone resorption. Has ability to cross mucous membranes. Has no significant effect on inositol phosphate accumulation.
TRUSTED_SUSTAINABILITY

Specifications

Species Salmon
Purification Method HPLC
Purity ≥98%
Content And Storage Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 month at −20°C.
Format Lyophilized
Formulation Sold on the basis of peptide content.
Product Type Calcitonin
Quantity 25 mg
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.