Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calmodulin, Bovine Brain, High Purity, Protein

Catalog No. 2086945MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-869-45MG 5 mg
20-869-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-869-45MG Supplier MilliporeSigma™ Supplier No. 2086945MG
Only null left
Add to Cart
Add to Cart
A ubiquitous Ca2+-binding protein that serves as a physiological effector of a wide range of biological processes. Calmodulin highly purified from bovine brain.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 30mM HEPES, 1mM CaCl2, 0.1mM DTT, pH 7.4.
Preparation Method Aqueous buffers or H2O. Please refer to the vial label for lot-specific reconstitution volume.
Purification Method SDS-PAGE
Quantity 5 mg
Immunogen Calmodulin
Storage Requirements Following reconstitution, aliquot and freeze (−20°C or −70°C). Aqueous stock solutions are stable for up to 2 months at −20°C or for up to 4 months at −70°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%, High Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.