Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from protein solution with 2mM EDTA. |
| Preparation Method | Reconstitute with distilled water. H2O (1mg/mL) |
| Purification Method | Native PAGE |
| Quantity | 500 μg |
| Immunogen | Calmodulin |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C or −70°C). Aqueous stock solutions are stable for up to 2 months at −20°C or for up to 4 months at −70°C. |
| Regulatory Status | RUO |
| Species | Bovine |
| Recombinant | Native |
| Form | Lyophilized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.