Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calpain-1, Human Erythrocytes, Protein

Catalog No. 208713500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-871-3500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-871-3500UG Supplier MilliporeSigma™ Supplier No. 208713500UG
Add to Cart
Add to Cart
Native calpain-1 from human erythrocytes. Ca2+-dependent heterodimeric cysteine proteinase with low Ca2+ requirement (EC50= 2μM).
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 20mM imidazole, 5mM β-mercaptoethanol, 1mM EDTA, 1mM EGTA, 30% glycerol, pH 6.8.
Purification Method Comparable to reference lot by SDS-PAGE
Quantity 500 μg
Immunogen Calpain-1
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.