Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Carcinoembryonic Antigen, Human Colon Adenocarcinoma Cell Line, Protein

Catalog No. 219369100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-936-9100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-936-9100UG Supplier MilliporeSigma™ Supplier No. 219369100UG
Add to Cart
Add to Cart
Native CEA from human colon adenocarcinoma cells. Found in tumors in adults. Presence indicates carcinogenic activity. Purified CEA has been the most widely-used general marker in determining and monitoring the status of malignant disease.

Specifications

Preparation Method Aqueous Buffers
Purification Method Single diffuse band by SDS-PAGE
Quantity 100 μg
Immunogen Carcinoembryonic Antigen
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.