Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cathepsin B, Human Liver, >95% by SDS-PAGE, Protein

Catalog No. 2193645U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-936-45U 5U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-936-45U Supplier MilliporeSigma™ Supplier No. 2193645U
Only null left
Add to Cart
Add to Cart
Native cathepsin B from human liver, purified by affinity chromatography. The most investigated enzyme of all lysosomal cysteine proteases. Upregulation and secretion of cathepsin B have been shown to occur in many types of tumors and to correlate positively with their invasive and metastatic capabilities. Cathepsin B is shown to facilitate tumor invasion by dissolving extracellular barriers.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50mM sodium acetate buffer, 1mM EDTA, pH 5.0.
Purification Method SDS-PAGE
Quantity 5U
Immunogen Cathepsin B
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.