Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ CBX8 Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710135MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-35MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-35MI Supplier MilliporeSigma™ Supplier No. 1710135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects CBX8 Clone: in Human samples, and it is validated for ChIP, Western Blotting

The family of Heterochromatin Proteins (Chromobox Homolog, CBX) are highly conserved adapter molecules, which have important functions in the cell nucleus. These functions include gene repression by heterochromatin formation, transcriptional activation, regulation of binding of cohesion complexes to centromere, sequesteration of genes to nuclear periphery, transcriptional arrest, maintenance of heterochromatin integrity, gene repression at single nucleosome level and gene repression by heterochromatization of euchromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX8
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-CBX8 (Rabbit Polyclonal). One vial containing 50μg affinity purified rabbit polyclonal in 72μL buffer containing Tris-citrate/phosphate buffer (pH 7 to 8) and 0.09% sodium azide, before the addition of glycerol to 30% (0.7mg/mL final). Normal Rabbit IgG. One vial containing 125μg of purified Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, HOXA2 Upstream. One vial containing 75μL of 5μM of each primer specific for human HOXA2 promoter region (chr7:27142948-27143085, hg19 build). FOR: AGG AAA GAT TTT GGT TGG GAA G; REV: AAA AAG AGG GAA AGG GAC AGA C
Gene Accession No. Q9HC52
Gene Symbols HPC3, PC3, Pc3, RC1, hPc3
Host Species Rabbit
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_065700
Includes Use ChIPAb+ CBX8 ChIP Validated rabbit polyclonal antibody for the detection of CBX8 also known as Chromobox protein homolog 8 and Polycomb 3 homolog.
Target Species Human
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.