Learn More
MilliporeSigma™ CDS10 Selector Valve
GSA_VA
Catalog No. 6003 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1/Pk.
Catalog No. Quantity
6003 1/Pk.
1 options
Catalog No. 6003 Supplier MilliporeSigma™ Supplier No. 6003
Add to Cart
Add to Cart
Simplifies operation and avoids the need for multiple T-fittings and valves
  • With sliding control for instant switching from concentration to diafiltration, or switching gas and liquid lines simultaneously
  • Made of Acetal, Buna-N and nylon material
  • Chemically sanitizable
  • Dimensions: 3 x 0.7 x 3.1 in. (7.5 x 2 x 8cm)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) switch the Amicon Stirred Cell between concentration and diafiltration modes
Material Acetal, Buna-N and nylon
Quantity 1/Pk.