Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Certistain™ Fuchsin Acid Dye

Catalog No. M1052310025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1052310025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1052310025 Supplier MilliporeSigma™ Supplier No. 1.05231.0025

missing translation for 'validMainframeMsg'

Fuchsin acid (C.I. 42685) for microscopy Certistain™. CAS 3244-88-0, pH 3-4 (10g/l, H2O, 20°C).

The CERTISTAIN™ dry dye Acid fuchsin (C.I. 42685) is used for the preparation of a staining solution that can be used for staining connective tissues in histological sections with trichrome staining acc. to Mallory and van Gieson.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 g
For Use With (Application) Prepares staining solution used for staining connective tissues in histological sections with trichrome staining
Color Green
Melting Point >130°C (decomposition)
pH pH: 3-4
Product Type Fuchsin Acid Dye
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.