Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Certistain™ New Fuchsin Dye

Catalog No. M1052260100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1052260100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1052260100 Supplier MilliporeSigma™ Supplier No. 1.05226.0100

New fuchsin (C.I. 42520) for microscopy Certistain™. CAS 3248-91-7, pH 3-4 (10g/l, H2O, 20°C).

The CERTISTAIN™ dry dye New fuchsin (C.I. 42520) is used for the preparation of a staining solution that can be used for staining of histological tissue sections or other samples of human origin according to Ziehl-Neelsen, Gram staining as well as in combination with Schiff’s reagent.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
For Use With (Application) Used for preparation of staining solution that can be used for staining of histological tissue sections
Product Type New Fuchsin Dye
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.