Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Certistain™ Rhodamine B Dye

Catalog No. M1075990100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1075990100 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1075990100 Supplier MilliporeSigma™ Supplier No. 1.07599.0100

missing translation for 'validMainframeMsg'

Rhodamine B (C.I.45170) for microscopy. CAS 81-88-9, pH 3-4 (10g/l, H2O, 20°C).

Rhodamine B (C.I. 45170) is a dry dye for fluorescence Tb staining of bacteriological smears and histological tissue sections of human origin.The mechanism is a non-specific binding of fluorescence dye on a substrate, similar to the classical Ziehl-Neelsen method, although the heating of the Rhodamine B color solution can be avoided. The dye is a CE certified IVD product and has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100g
For Use With (Application) Fluorescence Tb staining of bacteriological smears, histological tissue sections of human origin
Melting Point 199-201°C
Product Type Rhodamine B Dye
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.