Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ CHD1, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710533MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-105-33MI 10 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-105-33MI Supplier MilliporeSigma™ Supplier No. 1710533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects CHD1 Clone: in Human samples, and it is validated for ChIP, Western Blotting

Chromodomain-helicase-DNA-binding protein 1 (CHD1) is a multidomain chromatin-remodelling enzyme, which belongs to the SNF2/RAD54 helicase family. It contains an SWI/SNF-like helicase/ATPase domain, a DNA-binding domain, and two chromodomains. CHD1 is multifunctional protein, which has an important role in gene regulation. It interacts with various modified histones, such as H3K4Me2 and H3K4Me3, and maintains the open chromatin structure required for transcription. It also interacts with, or plays a role in the assembly of, several transcription-related complexes, including the histone acetylation complex SAGA, and the FACT and PAF complexes. Previous studies have suggested that CHD1 maintains the pluripotent state of embryonic stem cells, and is required for the reprogramming of somatic cells to the pluripotent state. CHD1 may play a role in the development of prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD1
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-CHD1 (Rabbit Polyclonal). One vial containing 7.0μg purified rabbit polyclonal in 50μL buffer of Tris-buffered Saline containing 0.1% BSA and 0.09% Sodium Azide, before the addition of 30% glycerol (0.14mg/mL final). Normal Rabbit IgG. One vial containing 125μg of purified Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, PSMC6. One vial containing 75μL of 5μM of each primer specific for human proteasome 26S subunit 6 (chr14:53174161+53174274 , hg19 build). Primer location selection based on ENCODE browser data at UCSC, see also reference 1. FOR: TCC GGC CCT GAG CTT GT; REV: CCT CCT CTA CTT CTT TTT CAT TTT CAC
Gene Accession No. O14646
Gene Symbols Chd1, Chd-1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human CHD1 (a.a. 1660-1710).
Quantity 10 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001261
Test Specificity Recognizes CHD1 in regions between a.a. 1660 to 1710.
Includes This ChIPAb+ Validated Antibody and Primer Set conveniently includes the antibody, matched IgG negative control antibody and set control PCR primers that detect a known positive locus.
Target Species Human
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.