Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Accutase XL Lyophilized Accutase

Catalog No. SCR104 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR104 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR104 Supplier MilliporeSigma™ Supplier No. SCR104
Add to Cart
Add to Cart

a lyophilized form of Accutase with extended stability used for the routine detachment of cells from standard tissue culture plasticware

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20
For Use With (Application) Cell culture and cell harvesting
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.