Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ AOF ITS Supplement (100x)

Catalog No. SCM054 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM054 10 mL
SCM055 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SCM054 Supplier MilliporeSigma™ Supplier No. SCM054
Only null left
Add to Cart
Add to Cart

an animal-origin-free product that contains human recombinant insulin, transferrin along with sodium selenite and is used for the serum-free culture of numerous cell types.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2-8°C
For Use With (Application) Stem cell culture, cell culture, cell stimulation, cell growth
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.