Learn More
MilliporeSigma™ Chemicon™ Dopaminergic Neuron Integration and Characterization Kit
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Applications | Neuron Integration and Characterization, Stem Cell Research |
| Description | Dopaminergic neuron integration and characterization kit |
| Quantity | 1 Kit |
| Content And Storage | -NeuN mouse monoclonal MAB377 -Dopa Decarboxylase rabbit polyclonal AB1569 -DAT rat monoclonal MAB369 -GFAP rabbit polyclonal AB5804 -ChAT goat polyclonal AB144P -Serotonin rat monoclonal MAB352 -Tyrosine Hydroxylase mouse monoclonal MAB5280 |
| Product Type | Dopaminergic neuron integration and characterization kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.