Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ EndoGRO™-MV Complete Culture Media Kit
GSA_VA

Catalog No. SCME004 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCME004 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCME004 Supplier MilliporeSigma™ Supplier No. SCME004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

a 5% serum formulation for culturing human microvascular endothelial cells, this media does not contain VEGF.

EndoGRO Media Formulations provide an optimal cell culture environment for many types of endothelial cells, including HUVEC, aortic endothelial cells and other human large vessel and microvascular endothelial cells. These media have been shown to grow endothelial cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. The media are packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge that helps prevent contamination. EndoGRO Media contain no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental results. Four different media formulations are available for various applications and cell culture requirements.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell culture
Quantity 500 mL
Content And Storage Basal medium: 2-8°C in dark; Supplement Mix: -20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.