Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Human Mesenchymal-LS Expansion Medium

Catalog No. SCM023 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM023 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM023 Supplier MilliporeSigma™ Supplier No. SCM023
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

a low FBS (2%) cell culture media for human mesenchymal stem cells (hMSCs) including those derived from human adipose, bone marrow and ES/iPS cells.

Human Mesenchymal-LS Expansion Medium provides a low serum (2%) cell culture environment for human mesenchymal stem cells including those derived from human adipose and bone marrow tissues and ES/iPS cells. This low serum formulation have been shown to grow cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. Human Mesenchymal-LS Expansion Medium is specific to human cells and should not be used to culture mouse or rat MSCs. The medium is packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge for added convenience. Human Mesecnhymal-LS Expansion Medium contains no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental result.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Quantity 500 mL
Content And Storage Basal Medium: 2 - 8°C; Supplement Kit: -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.