Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Human Mesenchymal-XF Expansion Medium

Catalog No. SCM045 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM045 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM045 Supplier MilliporeSigma™ Supplier No. SCM045
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xeno-free mesenchymal stem cell media

specially formulated for the rapid expansion of human mesenchymal stem cells isolated from a variety of sources, including adipose and bone marrow tissues.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Stem Cell Culture, Cell Culture
Quantity 500 mL
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.