Learn More
MilliporeSigma™ Chemicon™ Human OPC Spontaneous Differentiation Culture Media Kit
Catalog No. SCM106 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
SCM106 100 mL
1 options
Catalog No. SCM106 Supplier MilliporeSigma™ Supplier No. SCM106
Add to Cart
Add to Cart
a defined, serum-free medium that supports the differentiation of human ESC-derived OPCs.
Human OPC Spontaneous Differentiation Media is a defined, serum-free medium that supports the differentiation of human ESC-derived OPCs (Catalog No. SCR600) to mature oligodendrocytes through growth factor withdrawal. The media does not support the differentiation of primary oligodendrocyte progenitor cells isolated from brain tissues
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Stem Cell Culture
Quantity 100 mL