Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Human Recombinant TGF-beta2

Catalog No. PF0171UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PF0171UG 1 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF0171UG Supplier MilliporeSigma™ Supplier No. PF0171UG

Specifications

Content And Storage -20°C in undiluted aliquots for up to 6 months
Form Lyophilized
Quantity 1 μg
Source E. coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.