Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ LentiBrite™ GFP-LC3-II Enrichment Kit (Flow Cytometry)

Catalog No. 1710230 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-102-30 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-30 Supplier MilliporeSigma™ Supplier No. 1710230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

For Use With (Application) Flow Cytometry
Includes TagGFP2-LC3 Lentivirus: One vial containing 25μL of lentiviral particles at a minimum of 3 x 10E8 infectious units (IFU) per mL, Promoter: EF-1 (Elongation Factor-1), Autophagy Reagent A: One vial (lyophilized), 10x Enrichment Buffer: One vial containing 1 mL, 5x Assay Buffer: One bottle containing 3 mL
Quantity 1 Kit
Concentration Minimum of 3 x 10E^8 Infectious Units (IFU)/mL
Content And Storage Store TagGFP2-LC3 Lentivirus at -80°C. Store Autophagy Reagent A, 10x Enrichment Buffer, and 5x Assay Buffer at 2-8°C
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.