Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Leptin (Anti-Obesity Protein)

Catalog No. GF050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF050 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF050 Supplier MilliporeSigma™ Supplier No. GF050
Add to Cart
Add to Cart

Protein product of ob (obese) gene in mice and appears to be involved in appetite control

  • Expressed in adipose tissue
  • Regulates fat stores in body
  • Mice with the ob/ob genotype also develop form of diabetes similar to type II (non-insulin dependent) diabetes
  • Molecular weight: 16kd
  • Contains 147 amino acid residues
  • Lyophilized from 0.1% TFA
  • w/o Preservatives
  • Purity: >95% by SDS-PAGE and HPLC analyses
  • Source: E.coli
  • Speies: Mouse

Recommended Storage

Store dessicated below 0°C

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Quantity 1 mg
Source E.coli
Storage Requirements Store dessicated at 0°C
Product Type Leptin (Anti-Obesity Protein)
Species Mouse
Purity or Quality Grade >95% by SDS-PAGE and HPLC analyses
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.