Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Chemicon™ LT2 Immortalized Pancreatic Mesenchymal Cell Line
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Content And Storage | Store in vapor phase of liquid nitrogen |
| For Use With (Application) | Stem cell culture |
| Quantity | 5 x 105 Cells |
| Source | Human fetal pancreatic fibroblasts |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?