Learn More
MilliporeSigma™ Chemicon Macrophage Inflammatory Protein-1alpha Protein
Shop All MilliporeSigma ProductsDescription
- Involved in acute inflammatory state in recruitment and activation of polymorphonuclear leukocytes
- Recombinant
- Molecular weight: 7.8kd
- Contains 69 amino acid residues
- Source: E.coli
- Species: Rat
- Purity: >95% by SDS-PAGE and HPLC analysis
- Lyophilized powder
Recommended Storage
Store at -20°C
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 1 mg |
| Source | E.coli |
| Storage Requirements | Store dessicated at 0°C |
| Product Type | eptin (Anti-Obesity Protein) |
| Species | Mouse |
| Purity or Quality Grade | >95% by SDS-PAGE and HPLC analyses |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.