Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Mouse Neural Stem Cell Expansion Medium

Catalog No. SCM008 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM008 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM008 Supplier MilliporeSigma™ Supplier No. SCM008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

allows for the growth and proliferation of mouse neural stem cells.

The Mouse Neural Stem Cell Expansion Medium allows for the growth and proliferation of mouse neural stem cells. This product includes the Neural Stem Cell Basal Medium and the supplements bFGF, EGF, and Heparin. Withdrawal of the growth factors and heparin from the Neural Stem Cell Basal Medium will result in the spontaneous differentiation of mouse neural stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 500mL of Neural Stem Cell Basal Medium (Mfr. No. SCM003); Heparin, 1.5mg; EGF, 10μg; bFGF, 10μg. Store at -20°C
For Use With (Application) Stem cell culture
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.