Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Neurite Growth Protein L1

Catalog No. GF220 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF220 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF220 Supplier MilliporeSigma™ Supplier No. GF220

missing translation for 'validMainframeMsg'

a cell adhesion molecule of the Ig superfamily (IgCAM) plays a critical role in the formation of neuronal networks.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C for up to 6 months
For Use With (Application) Cell adhesion molecule of Ig superfamily (IgCAM) plays critical role in the formation of neuronal networks
Quantity 100 μg
Source Mammalian cells, HEK293 cell line
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.