Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Pancreatic Cell Culture Supplement

Catalog No. SCR015 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR015 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR015 Supplier MilliporeSigma™ Supplier No. SCR015

intended for the preparation of complete cell culture medium in conjunction with Pancreatic cell culture Medium (SCR016).

Pancreatic Cell Culture Supplement is intended for the preparation of complete cell culture medium in conjunction with Pancreatic Cell Culture Medium (SCR016). This complete medium is optimized for the culture of LT2 Immortalized Pancreatic Mesenchymal Cells (SCR013) and VIT1 Primary Pancreatic Mesenchymal Cells (SCR014).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20
For Use With (Application) Cell culture
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.