Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Rat ESGRO™ 10 million units/1mL

Catalog No. ESG2207 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ESG2207 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ESG2207 Supplier MilliporeSigma™ Supplier No. ESG2207
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Applications for Rat ESGRO include use as a reagent for the in vitro maintenance of rat ES cells. rtLIF is more effective in maintaining the undifferentiated phenotype of rat ES cells than similar concentrations of murine LIF.

Leukaemia inhibitory factor is a cytokine that induces macrophage differentiation. Neurotransmitters and neuropeptides, well known for their role in the communication between neurons, are also capable of activating monocytes and macrophages and inducing chemotaxis in immune cells. LIF signals through different receptors and transcription factors. LIF in conjunction with BMP2 acts in synergy on primary fetal neural progenitor cells to induce astrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Form Purified
For Use With (Application) Stem cell culture
Quantity 1 mL
Source E. coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.