Learn More
Description
- Signalling has been implicated in control of differentiation of stem cells
- Lyophilized from solution containing PBS and 1% CHAPS buffer
- Endotoxin level: <0.1ng/μg (1EU/μg)
- Source: Mouse L cells expressing full length Wnt3a
- Species: Mouse
- Molecular weight: 38-41kd
Recommended Storage
Store at -20°C
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C |
| Endotoxin Concentration | <0.1ng/μL of Wnt5a |
| Gene Symbol | WNT5A |
| Product Type | Wnt-3a Protein |
| Species | Mouse |
| Recombinant | Recombinant |
| Protein Family | Wnt protein family |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.