Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Recombinant Mouse Wnt-5a Protein

Catalog No. gf146 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF146 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF146 Supplier MilliporeSigma™ Supplier No. GF146
Only null left
Add to Cart
Add to Cart

Contains cysteine modified by palmitate essential for receptor binding and biological activity

  • Recombinant
  • Contains 352 amino acid protein containing 23 conserved cysteines
  • Contains N-terminal signal peptide
  • Protein is secreted into medium when expressed in cell culture
  • Purified from conditioned media using blue-Sepharose™, gel filtration and heparin affinity chromatography
  • Frozen liquid in PBS containing 1% Chaps
  • Concentration: 80-100ng/μL Endotoxin level: 0.1ng/μg of Wnt5a

Recommended Storage

Store dessicated at -20°C

TRUSTED_SUSTAINABILITY

Specifications

Concentration 80-100ng/µL
For Use With (Application) Cell Culture
Purification Method Purified from conditioned media using blue-Sepharose™, gel filtration and heparin affinity chromatography
Quantity 10 μL
Storage Requirements Store frozen at -20°C
Endotoxin Concentration <0.1ng/μL of Wnt5a
Gene Symbol WNT5A
Product Type Wnt-5a Protein
Species Mouse
Recombinant Recombinant
Protein Family Wnt protein family
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.