Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ ReNcell™ CX Human Neural Progenitor Cell Line and Culture Media Kit

Catalog No. SCC009 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC009 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC009 Supplier MilliporeSigma™ Supplier No. SCC009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

contains human neural progenitor cells, optimized maintenance media and freezing media for culture of human neural stem cells.

ReNcell CX Kit contains human neural progenitor cells, optimized maintenance media and freezing media for culture of human neural stem cells. ReNcell CX Immortalized Cells (SCC007): ReNcell CX is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons and glial cells. ReNcell CX was derived from the cortical region of human fetal brain. Immortalized by retroviral transduction with the c-myc oncogene, this cell line grows rapidly as a monolayer on laminin with a doubling time of 20-30 hours. Karyotype analyses indicate that the ReNcell CX retains a normal diploid karyotype in culture even after prolonged passage (>45 passages). ReNcell CX was developed by the ReNeuron Group plc, a biotech company that specializes in using human somatic stem cells for therapeutics. ReNcell CX may be used for a variety of research applications such as studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions. ReNcell CX cells have been obtained in a legal and ethical manner, compliant with current local informed consent procedures. ReNcell NSC Maintenance Medium (SCM005): ReNcell Neural Stem Cell (NSC) Maintenance Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of ReNcell immortalized human neural progenitor cells. When used in conjunction with FGF and EGF, the maintenance medium will allow for the proliferation of ReNcell immortalized VM and CX neural stem cells. Withdrawal of the growth factors from ReNcell NSC Maintenance Medium will result in the spontaneous differentiation of ReNcell immortalized neural progenitor cells. ReNcell NSC Freezing Medium (SCM007): ReNcell NSC Freezing Medium is qualified for use with ReNcell immortalized human neural progenitor cell lines, CX (CHEMICON Catalog No. SCC007) and VM (CHEMICON Catalog No. SCC008) cultured in serum-free conditions with ReNcell NSC Maintenance Medium (CHEMICON Catalog No. SCM005). The optimized formulation allows for consistent cryopreservation and high viability upon thawing and plating.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ReNcell Neural Stem Cell Freezing Medium (Mfr. No. SCM007); ReNcell CX Immortalized Cell Line (Mfr. No. SCC007); ReNcell NSC Maintenance Media (Mfr. No. SCM005)
For Use With (Application) Stem cell culture
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.