Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ ReNcell™ VM Human Neural Progenitor Cell Line

Catalog No. SCC008 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC008 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC008 Supplier MilliporeSigma™ Supplier No. SCC008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons and glial cells.

ReNcell VM is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons and glial cells. ReNcell VM was derived from the ventral mesencephalon region of human fetal brain. Immortalized by retroviral transduction with the v-myc oncogene, this cell line grows rapidly as a monolayer on laminin with a doubling time of 20-30 hours. Karyotype analyses indicate that the ReNcell VM retains a normal diploid karyotype in culture even after prolonged passage (>45 passages). ReNcell VM was developed by the ReNeuron Group plc, a biotech company that specializes in using human somatic stem cells for therapeutics. In experiments performed by the ReNeuron Group plc, ReNcell VM can be differentiated in vitro to a high level of human dopaminergic neurons. Neurons differentiated from ReNcell VM have furthermore been shown to be electophysiologically active. ReNcell VM may be used for a variety of research applications such as studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions. ReNcell VM cells have been obtained in a legal and ethical manner, compliant with current local informed consent procedures.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Stem cell culture
Quantity 1 Ea.
Source Human ventral mesencephalon brain tissue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.