Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ ReNcell™ VM Immortalized Cell Kit

Catalog No. SCC010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC010 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC010 Supplier MilliporeSigma™ Supplier No. SCC010
Add to Cart
Add to Cart

contains human neural progenitor cells, optimized maintenance media and freezing media for culture of human neural stem cells.

ReNcell VM Kit contains human neural progenitor cells, optimized maintenance media and freezing media for culture of human neural stem cells. ReNcell VM Immortalized cells (SCC008): ReNcell VM is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons and glial cells. ReNcell VM was derived from the ventral mesencephalon region of human fetal brain. Immortalized by retroviral transduction with the v-myc oncogene, this cell line grows rapidly as a monolayer on laminin with a doubling time of 20-30 hours. Karyotype analyses indicate that the ReNcell VM retains a normal diploid karyotype in culture even after prolonged passage (>45 passages). ReNcell VM was developed by the ReNeuron Group plc, a biotech company that specializes in using human somatic stem cells for therapeutics. In experiments performed by the ReNeuron Group plc, ReNcell VM can be differentiated in vitro to a high level of human dopaminergic neurons. Neurons differentiated from ReNcell VM have furthermore been shown to be electophysiologically active. ReNcell VM may be used for a variety of research applications such as studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions. ReNcell VM cells have been obtained in a legal and ethical manner, compliant with current local informed consent procedures. ReNcell NSC Maintenance Medium (SCM005): ReNcell Neural Stem Cell (NSC) Maintenance Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of ReNcell immortalized human neural progenitor cells. When used in conjunction with FGF and EGF, the maintenance medium will allow for the proliferation of ReNcell immortalized VM and CX neural stem cells. Withdrawal of the growth factors from ReNcell NSC Maintenance Medium will result in the spontaneous differentiation of ReNcell immortalized neural progenitor cells. ReNcell NSC Freezing Medium (SCM007): ReNcell NSC Freezing Medium is qualified for use with ReNcell immortalized human neural progenitor cell lines, CX (CHEMICON Catalog No. SCC007) and VM (CHEMICON Catalog No. SCC008) cultured in serum-free conditions with ReNcell NSC Maintenance Medium (CHEMICON Catalog No. SCM005). The optimized formulation allows for consistent cryopreservation and high viability upon thawing and plating.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ReNcell VM Immortalized Cell LIne (Mfr.No. SCC008); ReNcell Neural Stem Cell Freezing Medium (Mfr. No. SCM007); ReNcell NSC Maintenance Media (Mfr . No. SCM005)
For Use With (Application) Stem cell culture
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.