Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ ChIPAb+™ HDAC1 Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710199MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-99MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-99MI Supplier MilliporeSigma™ Supplier No. 1710199
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifically detects ChIPAb+™ HDAC1 Clone: in Canine, Hamster, Human, Mouse, Rat samples, and it is validated for ChIP, Immunocytochemistry, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ChIPAb+™ HDAC1
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-HDAC1 (rabbit polyclonal). One vial containing 50μg of Anti-HDAC1 IgG in PBS with 1mg/mL BSA and ≤0.1% sodium azide before the addition of glycerol to 30%. Concentration: 0.7mg/mL Normal Rabbit IgG. One vial containing 125μg of rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers p21. One vial containing 75μL of 5μM of each primer specific for a region of the human p21 (WAF1/CIP1/CDKN1A) promoter. FOR: CCC ACA GCA GAG GAG AAA GAA; REV: CTG GAA ATC TCT GCC CAG ACA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 467-482 of human HDAC1.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_032254
Includes This ChIPAb+ HDAC1 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Canine, Hamster, Human, Mouse, Rat
Content And Storage -20°C for one year from date of shipment; Aliquot to avoid repeated freezing and thawing; For maximum recovery of product centrifuge the original vial after thawing and prior to removing the cap
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.