Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ ChIPAb+™ Phospho-Histone H3 (Thr3)α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710141MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-41MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-41MI Supplier MilliporeSigma™ Supplier No. 1710141
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifically detects ChIPAb+™ Phospho-Histone H3 (Thr3)α Clone: in Human samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ChIPAb+™ Phospho-Histone H3 (Thr3)α
Applications ChIP Assay, Dot Blot, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Phospho-Histone H3 (Thr3) (rabbit monoclonal). One vial containing 50μL of cultured supernantant in 0.05% sodium azide. Negative Control Supernatant. One vial containing 100μL of cultured supernatant in 0.05% sodium azide. Control Primers, human GAPDH promoter. One vial containing 75μL of 5μM of each primer specific for human GAPDH. FOR: TAC TAG CGG TTT TAC GGG CG; REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. Q16695
Gene Symbols HIST3H3, H3FT, MGC126886, H3t, MGC126888, H3.4, H3T, H3/g, H3/t
Host Species Rabbit
Immunogen Peptide containing the sequence [RTtrimKQ] in which lysine 4 is trimethylated on human histone H3
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Test Specificity Histone H3 phosphorylated at Threonine 3; cross-reactivity with phospho-histone H3 Thr22 detected in some applications
Includes This ChIPAb+ Phospho-Histone H3 (Thr3) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Culture Supernatant
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.