Learn More
MilliporeSigma™ Chorionic Gonadotropin, Human Urine, Iodination Grade, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | Lyophilized solid containing 25mM NH4HCO3. |
| Preparation Method | H2O |
| Quantity | 100 μg |
| Immunogen | Chorionic Gonadotropin |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Solid |
| Purity or Quality Grade | Iodination Grade |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.