Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Coagulation Factor V, Human Plasma, Protein

Catalog No. 219365100UG Shop All MilliporeSigma Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-936-5100UG 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 21-936-5100UG missing translation for 'mfr' MilliporeSigma™ missing translation for 'supplierNo' 219365100UG
Add to Cart
Add to Cart
Native coagulation factor V from human plasma. Thrombin cleaves Factor V to yield activated Factor Va, which exhibits 50-fold greater activity than the precursor molecule. Factor Va, Ca2+, and membrane phospholipids are cofactors that form a 'prothrombinase' complex with Factor X, enabling its proteolytic activation of prothrombin to thrombin.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50% glycerol/H2O (v/v).
Purification Method SDS-PAGE
Quantity 100μg
Immunogen Coagulation Factor V
Storage Requirements Following initial thaw, aliquot and freeze (−20°C). Do not store below −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.