Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 100mM NaCl, 20mM Tris-HCl, 1mM benzamidine, pH 7.4. |
| Purification Method | Single band by SDS-PAGE |
| Quantity | 100 U |
| Immunogen | Coagulation Factor X |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.